missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxybenzoate, 99%

Product Code. 11449263
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449263 10 g 10g
11459263 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449263 Supplier Thermo Scientific Alfa Aesar Supplier No. A11735.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-18-8 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-18-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016471
InChI Key MOZDKDIOPSPTBH-UHFFFAOYSA-N
Synonym benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
PubChem CID 7180
ChEBI CHEBI:34571
IUPAC Name benzyl 4-hydroxybenzoate
SMILES OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 110°C to 114°C
Quantity 10 g
Beilstein 2115995
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl 4-hydroxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.