missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Cyano-4-deoxy-2,3-O-[(1S,2S)-1,2-dimethoxy-1,2-dimethyl-1,2-ethanediyl]-Beta-D-arabinopyranoside, TRC

Product Code. 30329469
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329469 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329469 Supplier Toronto Research Chemicals Supplier No. TRCB23378010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 4-Cyano-4-deoxy-2,3-O-[(1S,2S)-1,2-dimethoxy-1,2-dimethyl-1,2-ethanediyl]-beta-D-arabinopyranoside
CAS 1084896-40-1
Molecular Formula C19H25NO6
Quantity 10 mg
Synonym Phenylmethyl 4-Cyano-4-deoxy-2,3-O-[(1S,2S)-1,2-dimethoxy-1,2-dimethyl-1,2-ethanediyl]-beta-D-arabinopyranoside
InChI Formula InChI=1S/C19H25NO6/c1-18(21-3)19(2,22-4)26-16-15(25-18)14(10-20)12-24-17(16)23-11-13-8-6-5-7-9-13/h5-9,14-17H,11-12H2,1-4H3/t14-,15-,16+,17-,18+,19+/m1/s1
SMILES CO[C@@]1(C)O[C@@H]2[C@H](OCc3ccccc3)OC[C@@H](C#N)[C@H]2O[C@]1(C)OC
IUPAC Name (2S,3S,4aS,5S,8R,8aR)-2,3-dimethoxy-2,3-dimethyl-5-phenylmethoxy-5,7,8,8a-tetrahydro-4aH-pyrano[3,4-b][1,4]dioxine-8-carbonitrile
Molecular Weight (g/mol) 363.4
Formula Weight 363.1682
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.