missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-chlorophenyl ketone, 98%

Product Code. 11498516
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498516 25 g 25g
11488516 5 g 5g
11408526 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498516 Supplier Thermo Scientific Alfa Aesar Supplier No. A15959.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1889-71-0 | C14H11ClO | 230.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1889-71-0
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00016342
InChI Key DXVALSKCLLBZEB-UHFFFAOYSA-N
Synonym benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by
PubChem CID 233840
IUPAC Name 1-(4-chlorophenyl)-2-phenylethanone
SMILES ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1

Specifications

Melting Point 103°C to 107°C
Quantity 25 g
Beilstein 976443
Formula Weight 230.69
Percent Purity 98%
Chemical Name or Material Benzyl 4-chlorophenyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.