missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 3-hydroxyphenylacetate, 98%

Product Code. 11450548
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450548 5 g 5g
11440548 1 g 1g
11460548 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450548 Supplier Thermo Scientific Alfa Aesar Supplier No. B24679.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 295320-25-1 | C15H14O3 | 242.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295320-25-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.27
MDL Number MFCD04039768
InChI Key ALFOBMRIXXPJLQ-UHFFFAOYSA-N
Synonym benzyl 3-hydroxyphenylacetate, benzyl 2-3-hydroxyphenyl acetate, acmc-20alpo, benzyl-3-hydroxyphenylacetate, 3-hydroxyphenylacetic acid benzyl ester, phenylmethyl 2-3-hydroxyphenyl acetate, 3-hydroxy-phenyl-acetic acid benzyl ester
PubChem CID 7016416
IUPAC Name benzyl 2-(3-hydroxyphenyl)acetate
SMILES OC1=CC=CC(CC(=O)OCC2=CC=CC=C2)=C1

Specifications

Melting Point 79°C to 81°C
Quantity 5 g
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Benzyl 3-hydroxyphenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.