missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-Naphthyl Ether, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Benzyl 2-Naphthyl Ether
Specifications
Specifications
| Chemical Name or Material | Benzyl 2-Naphthyl Ether |
| CAS | 613-62-7 |
| Analyte or Component Names | Benzyl 2-Naphthyl Ether |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H14 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Naphthalene, 2-(phenylmethoxy)-, Naphthalene, 2-(benzyloxy)- (6CI,7CI,8CI), 2-(Phenylmethoxy)naphthalene, 2-(Benzyloxy)naphthalene, BON, BON (ether), Benzyl 2-naphthyl ether, Benzyl β-naphthyl ether, β-(Benzyloxy)naphthalene, β-Naphthol benzyl ether, β-Naphthyl benzyl ether |
| InChI Formula | InChI=1S/C17H14O/c1-2-6-14(7-3-1)13-18-17-11-10-15-8-4-5-9-16(15)12-17/h1-12H,13H2 |
| SMILES | C(Oc1ccc2ccccc2c1)c3ccccc3 |
| IUPAC Name | 2-benzyloxynaphthalene |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction