missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-naphthyl ether, 99%, Thermo Scientific™

Product Code. 11430457
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11430457

Brand: Thermo Scientific Alfa Aesar B21796.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-62-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.298
MDL Number MFCD00089092
InChI Key WLTCCDHHWYAMCG-UHFFFAOYSA-N
Synonym benzyl 2-naphthyl ether, 2-benzyloxy naphthalene, 2-benzyloxynaphthalene, naphthalene, 2-phenylmethoxy, 2-phenylmethoxy naphthalene, 2-phenylmethoxy-naphthalene, 2-benzyloxy-naphthalene, 2-phenylmethoxy naphthalenen, benzyl2-naphthylether, b-naphthylbenzyl ether
PubChem CID 123080
IUPAC Name 2-phenylmethoxynaphthalene
SMILES C1=CC=C(C=C1)COC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 96°C to 98°C
Quantity 500 g
Beilstein 2049922
Formula Weight 234.3
Percent Purity 99%
Chemical Name or Material Benzyl 2-naphthyl ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.