missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Chloroethyl Ether, TRC

Product Code. 30345825
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30345825 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345825 Supplier Toronto Research Chemicals Supplier No. TRCB2336851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 2-Chloroethyl Ether
CAS 17229-17-3
Molecular Formula C9H11ClO
Quantity 1 g
Synonym Benzyl 2-chloroethyl Ether, 1-(Benzyloxy)-2-chloroethane, 2-Benzyloxy-1-chloroethane, 2-Phenylmethyloxyethyl Chloride, Benzyl 2-Chloroethyl Ether, NSC 11269, alpha-(2-Chloroethoxy)toluene
InChI Formula InChI=1S/C9H11ClO/c10-6-7-11-8-9-4-2-1-3-5-9/h1-5H,6-8H2
SMILES ClCCOCc1ccccc1
IUPAC Name 2-chloroethoxymethylbenzene
Molecular Weight (g/mol) 170.64
Formula Weight 170.0498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.