missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoethyl ether, 97%

Product Code. 10187343 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10187343 5 g 5g
10722485 1 g 1g
10390204 25 g 25g
11994701 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10187343 Supplier Thermo Scientific Acros Supplier No. 388870050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1462-37-9 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1462-37-9
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD01321307
InChI Key FWOHDAGPWDEWIB-UHFFFAOYSA-N
Synonym benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene
PubChem CID 73833
IUPAC Name 2-bromoethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.3600g/mL
Boiling Point 254.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OCH2CH2Br
Quantity 5 g
Specific Gravity 1.36
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material Benzyl 2-bromoethyl ether
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 2-bromoethyl ether
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.