missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,3-O-[(1S,2S)-1,2-Dimethoxy-1,2-dimethyl-1,2-ethanediyl]-4-nitrobenzoyl-Alpha-L-xylopyranoside, TRC

Product Code. 30343053
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343053 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30343053 Supplier Toronto Research Chemicals Supplier No. TRCB27613050MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl 2,3-O-[(1S,2S)-1,2-Dimethoxy-1,2-dimethyl-1,2-ethanediyl]-4-nitrobenzoyl-alpha-L-xylopyranoside
CAS 1084896-42-3
Molecular Formula C25H29NO10
Quantity 50 mg
Synonym Phenylmethyl 2,3-O-[(1S,2S)-1,2-Dimethoxy-1,2-dimethyl-1,2-ethanediyl]-4-nitrobenzoyl-alpha-L-xylopyranoside
InChI Formula InChI=1S/C25H29NO10/c1-24(30-3)25(2,31-4)36-21-20(35-24)19(15-33-23(21)32-14-16-8-6-5-7-9-16)34-22(27)17-10-12-18(13-11-17)26(28)29/h5-13,19-21,23H,14-15H2,1-4H3/t19-,20+,21-,23+,24-,25-/m0/s1
SMILES CO[C@@]1(C)O[C@@H]2[C@H](CO[C@@H](OCc3ccccc3)[C@H]2O[C@]1(C)OC)OC(=O)c4ccc(cc4)[N+](=O)[O-]
IUPAC Name [(2S,3S,4aS,5R,8S,8aR)-2,3-dimethoxy-2,3-dimethyl-5-phenylmethoxy-5,7,8,8a-tetrahydro-4aH-pyrano[3,4-b][1,4]dioxin-8-yl] 4-nitrobenzoate
Molecular Weight (g/mol) 503.5
Formula Weight 503.1791
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.