missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5-dimethyl-n-octylammonium Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzyldimethyloctylammonium D5 (phenyl D5) Chloride |
| Molecular Formula | C17 D5 H25 N . Cl |
| Quantity | 0.05 g |
| Synonym | Benzyldimethyloctylammonium D5 (phenyl D5) Chloride, Benzyl-2,3,4,5,6-d5-dimethyl-n-octylammonium Chloride |
| InChI Formula | InChI=1 S/C17H30N.ClH/c1-4-5-6-7-8-12-15-18(2,3)16-17-13-10-9-11-14-17;/h9-11,13-14 H,4-8,12,15-16H2,1-3H3;1 H/q+1;/p-1/i9D,10 D,11 D,13 D,14 D; |
| SMILES | [Cl-].[2 H]c1c([2 H])c([2 H])c(C[N+](C)(C)CCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | dimethyl-octyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]azanium;chloride |
| Molecular Weight (g/mol) | 288.9106 |
| Formula Weight | 288.2381 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction