missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl-2,3,4,5,6-d5-dimethyl-n-decylammonium Chloride, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Benzyldimethyldecylammonium D5 (phenyl D5) Chloride |
| Molecular Formula | C19 D5 H29 N . Cl |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C19H34N.ClH/c1-4-5-6-7-8-9-10-14-17-20(2,3)18-19-15-12-11-13-16-19;/h11-13,15-16 H,4-10,14,17-18H2,1-3H3;1 H/q+1;/p-1/i11D,12 D,13 D,15 D,16 D; |
| SMILES | [Cl-].[2 H]c1c([2 H])c([2 H])c(C[N+](C)(C)CCCCCCCCCC)c([2 H])c1[2 H] |
| IUPAC Name | butyl-dimethyl-[(2,3,4,5,6-pentadeuteriophenyl)methyl]ammonium;chloride |
| Molecular Weight (g/mol) | 316.96 |
| Formula Weight | 316.2694 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction