missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,2,2-trichloroacetimidate, 98%

Product Code. 10299413 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299413 100 g Glass bottle 100g
10601822 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10299413 Supplier Thermo Scientific Acros Supplier No. 351601000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81927-55-1 | C9H8Cl3NO | 252.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81927-55-1
Molecular Formula C9H8Cl3NO
Molecular Weight (g/mol) 252.52
MDL Number MFCD00000805
InChI Key HUZCTWYDQIQZPM-UHFFFAOYSA-N
Synonym benzyl 2,2,2-trichloroacetimidate, benzyl trichloroacetimidate, benzyltrichloroacetimidate, trichloroacetimidic acid benzyl ester, 2,2,2-trichloro-1-phenylmethoxy ethanimine, ethanimidic acid, 2,2,2-trichloro-, phenylmethyl ester, benzyl 2,2,2-trichloroethanecarboximidate, benzyl2,2,2-trichloroacetimidate, acmc-209pmn, benzyl trichloracetimidate
PubChem CID 144968
IUPAC Name benzyl 2,2,2-trichloroethanimidate
SMILES ClC(Cl)(Cl)C(=N)OCC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3590g/mL
Boiling Point 109°C to 113°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CCl3C(=NH)OCH2C6H5
Refractive Index 1.5435 to 1.5455
Quantity 100 g
Specific Gravity 1.359
Solubility Information Solubility in water: reacts with water
Formula Weight 252.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzyl 2, 2, 2-trichloroacetimidate
Show More Show Less
compliance-icons
Product Identifier
  • Benzyl 2,2,2-trichloroacetimidate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.