Learn More
Benzyl 2,2,2-trichloroacetimidate, 98%
Description
Benzyl 2,2,2-trichloroacetimidate is used for the acid-catalyzed benzylation of hydroxy groups. It acts as reagent during the synthesis of funiculosin dimethyl ether and (S)-3-(benzyloxy)-2-methylpropanal. It can also be used for mild esterification of carboxylic acids in the presence of a catalytic amount of BF3 etherate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl 2,2,2-trichloroacetimidate is used for the acid-catalyzed benzylation of hydroxy groups. It acts as reagent during the synthesis of funiculosin dimethyl ether and (S)-3-(benzyloxy)-2-methylpropanal. It can also be used for mild esterification of carboxylic acids in the presence of a catalytic amount of BF3 etherate.
Solubility
Miscible with cyclohexane/dichloromethane.
Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Density | 1.356 |
| Boiling Point | 110°C to 114°C (0.5 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.545 |
| Quantity | 5 g |
| Beilstein | 2525375 |
| Sensitivity | Moisture sensitive |
| Synonym | benzyl 2,2,2-trichloroacetimidate, benzyl trichloroacetimidate, benzyltrichloroacetimidate, trichloroacetimidic acid benzyl ester, 2,2,2-trichloro-1-phenylmethoxy ethanimine, ethanimidic acid, 2,2,2-trichloro-, phenylmethyl ester, benzyl 2,2,2-trichloroethanecarboximidate, benzyl2,2,2-trichloroacetimidate, acmc-209pmn, benzyl trichloracetimidate |
| Solubility Information | Miscible with cyclohexane/dichloromethane. |
| Formula Weight | 252.53 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.