missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine N-Oxide, TRC

Product Code. 17944505
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17944505 5 mg 5mg
17972665 25 mg 25mg
17934395 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17944505 Supplier Toronto Research Chemicals Supplier No. TRCB2099605MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36504-71-9
Molecular Formula C19H23N3O2
Molecular Weight (g/mol) 325.4
Synonym N,N-Dimethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine;
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine oxide
SMILES CN(CCCOC1=NN(CC2=CC=CC=C2)C3=CC=CC=C31)(C)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H23N3O2/c1-22(2,23)13-8-14-24-19-17-11-6-7-12-18(17)21(20-19)15-16-9-4-3-5-10-16/h3-7,9-12H,8,13-15H2,1-2H3
Formula Weight 325.179
Percent Purity >95%
Chemical Name or Material Benzydamine N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.