missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine Hydrochloride, TRC

Product Code. 17923845
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17923845 10 g 10g
17914025 50 mg 50mg
17945055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17923845 Supplier Toronto Research Chemicals Supplier No. TRCB20995010G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-69-4
Molecular Formula C19 H23 N3 O . Cl H
Molecular Weight (g/mol) 345.87
Synonym Benzydamine Hydrochloride, N,N-Dimethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine hydrochloride, N,N-Dimethyl-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine monohydrochloride, 1-Benzyl-3-[3-(dimethylamino)propoxy]-1H-indazole monohydrochloride, 1-Benzyl-3-(3-dimethylaminopropoxy)-1H-indazole chloride, 1-Benzyl-3-[3-(dimethylamino)propoxy]-1H-indazole hydrochloride, 1-Benzyl-3-γ-dimethylaminopropoxy-1H-indazole hydrochloride, AF 864, Afloben, Alcidol, Algiflog, Andolex, Benalgin, Benciflam, Bendaminol, Benflogin, Benzidamine hydrochloride, Benzindamine hydrochloride, Benzydamine hydrochloride, Benzyrin, Difflam, Difflam Cream, Difflam Oral Rinse, Dorinamin, Enzamin, Imotryl, Ririlim, Riripen, Salyzoron, Saniflor, Tamas, Tantum, Verax
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CCCOc1nn(Cc2ccccc2)c3ccccc13

Specifications

Quantity 10 g
InChI Formula InChI=1S/C19H23N3O.ClH/c1-21(2)13-8-14-23-19-17-11-6-7-12-18(17)22(20-19)15-16-9-4-3-5-10-16;/h3-7,9-12H,8,13-15H2,1-2H3;1H
Formula Weight 345.1608
Percent Purity >95%
Chemical Name or Material Benzydamine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.