missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine hydrochloride, MedChemExpress

Product Code. 16427806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16427806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16427806 Supplier Medchem Express Supplier No. HY30235A
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzydamine hydrochloride is a locally-acting nonsteroidal anti-inflammatory drug with local anaesthetic and analgesic properties; selectively binds to prostaglandin synthetase and has notable in vitro antibacterial activity.

Benzydamine hydrochloride is a locally-acting nonsteroidal anti-inflammatory drug with local anaesthetic and analgesic properties

Specifications

Chemical Name or Material Benzydamine hydrochloride
Physical Form Solid
CAS 132-69-4
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H24ClN3O
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 41 mg/mL (118.54 mM)
SMILES CN(C)CCCOC1=NN(CC2=CC=CC=C2)C3=C1C=CC=C3.[H]Cl
Molecular Weight (g/mol) 345.87
Formula Weight 345.87
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.