missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benztropine Mesylate, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Specifications
Specifications
| Quantity | 25 mg |
| IUPAC Name | (1R,5R)-3-(diphenylmethoxy)-8-methyl-8-azabicyclo[3.2.1]octane; methanesulfonic acid |
| Formula Weight | 403.5 |
| Physical Form | Solid |
| Chemical Name or Material | Benztropine Mesylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction