missing translation for 'onlineSavingsMsg'
Learn More

Benzthiazide, MedChemExpress

Product Code. 16478756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478756 100 mg 100mg
16442276 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478756 Supplier Medchem Express Supplier No. HYB1424
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Benzthiazide is a long-acting diuretic and a hypertension agent. Benzthiazide is an inhibitor of carbonic anhydrase 9 (CA9), with Kis of 8.0, 8.8 and 10 nM for CA9, CA2 and CA1, respectively. Benzthiazide also suppresses proliferation of cancer cells.

Benzthiazide is a long-acting diuretic and a hypertension agent. Benzthiazide is an inhibitor of carbonic anhydrase 9 (CA9), with Kis of 8.0, 8.8 and 10 nM for CA9, CA2 and CA1, respectively. Benzthiazide also suppresses proliferation of cancer cells.

Specifications

Chemical Name or Material Benzthiazide
CAS 91-33-8
Color White
Purity Grade Notes Research
Molecular Formula C15H14ClN3O4S3
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (115.76 mM; Need ultrasonic)
SMILES O=S(C1=C(Cl)C=C(C2=C1)NC(CSCC3=CC=CC=C3)=NS2(=O)=O)(N)=O
Molecular Weight (g/mol) 431.94
Formula Weight 431.94
Percent Purity 98.9%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.