missing translation for 'onlineSavingsMsg'
Learn More

Benzoylnitromethane, 98%

Product Code. 10707963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10707963 5 g Glass bottle 5g
10631214 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10707963 Supplier Thermo Scientific Acros Supplier No. 372790050
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 614-21-1 | C8H7NO3 | 165.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-21-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00010218
InChI Key JTWHVBNYYWFXSI-UHFFFAOYSA-N
Synonym benzoylnitromethane, ethanone, 2-nitro-1-phenyl, alpha-nitroacetophenone, 2-nitro-1-phenylethanon, nitroacetophenone, alpha-nitro acetophenone, acmc-1bdpo, acetophenone, 2-nitro, nitromethyl phenyl ketone, .alpha.-nitroacetophenone
PubChem CID 94833
IUPAC Name 2-nitro-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 101°C to 106°C
Color Brown to Yellow or Colorless
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(O)CH2NO2
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in acetone,halogenated solvents,thf and,alcohols
Formula Weight 165.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Benzoylnitromethane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.