missing translation for 'onlineSavingsMsg'
Learn More

(Benzoylmethylene)triphenylphosphorane, 98+%

Product Code. 11404833
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404833 100 g 100g
11484823 5 g 5g
11494823 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404833 Supplier Thermo Scientific Alfa Aesar Supplier No. A12184.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 859-65-4 | C26H21OP | 380.43 g/mol

It is used in organic synthesis, wittig reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in organic synthesis, wittig reagent.

Solubility
Soluble in Methanol (almost transparency).

Notes
Air sensitive. Store in refrigerator. Store away from oxidizing agent, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 859-65-4
Molecular Formula C26H21OP
Molecular Weight (g/mol) 380.43
MDL Number MFCD00014088
InChI Key MZRSAJZDYIISJW-UHFFFAOYSA-N
Synonym benzoylmethylene triphenylphosphorane, 2-triphenylphosphoranylidene acetophenone, 1-phenyl-2-triphenylphosphoranylidene ethanone, phenacylidenetriphenyl phosphorane, phenylcarbonylmethylene triphenylphosphorane, acetophenone, 2-triphenylphosphoranylidene, phenacylidenetriphenylphosphorane, triphenylphosphinebenzoylmethylene, ethanone, 1-phenyl-2-triphenylphosphoranylidene
PubChem CID 70073
IUPAC Name 1-phenyl-2-(triphenyl-$l^{5}-phosphanylidene)ethanone
SMILES O=C(C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 183°C to 186°C
Quantity 100 g
Beilstein 620285
Sensitivity Air sensitive
Solubility Information Soluble in Methanol (almost transparency).
Formula Weight 380.43
Percent Purity ≥98%
Chemical Name or Material (Benzoylmethylene)triphenylphosphorane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.