missing translation for 'onlineSavingsMsg'
Learn More

Benzoylhypaconine, MedChemExpress

Product Code. 16499128
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499128 10 mg 10mg
16451538 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16499128 Supplier Medchem Express Supplier No. HYN0850

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Benzoylhypaconine (Benzoylhypacoitine) is a monoester Aconitum alkaloid, is the main pharmacologic and toxic component.

Benzoylhypaconine (Benzoylhypacoitine) is a monoester Aconitum alkaloid, is the main pharmacologic and toxic component.

Specifications

Chemical Name or Material Benzoylhypaconine
CAS 63238-66-4
Color White
Purity Grade Notes Research
Molecular Formula C31H43NO9
Quantity 10 mg
Synonym Benzoylhypacoitine
Solubility Information DMSO : 100 mg/mL (174.32 mM; Need ultrasonic)
SMILES CO[C@@H]1C2([C@@](C[C@@]3(O)[C@@H]4OC(C5=CC=CC=C5)=O)([H])[C@@]4([H])[C@](O)([C@@H](O)[C@@H]3OC)C6C2N(C)C7)[C@@]([C@H]6OC)([H])[C@@]7(COC)CC1
Molecular Weight (g/mol) 573.67
Formula Weight 573.67
Percent Purity 98.5%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.