missing translation for 'onlineSavingsMsg'
Learn More

Benzoylformic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4509384
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
611-73-4
C8H6O3
150.13
MFCD00002575
FAQJJMHZNSSFSM-UHFFFAOYSA-N
benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
11915
CHEBI:18280
2-oxo-2-phenylacetic acid
OC(=O)C(=O)C1=CC=CC=C1
This item is not returnable. View return policy
611-73-4
C8H6O3
150.13
MFCD00002575
FAQJJMHZNSSFSM-UHFFFAOYSA-N
benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
11915
CHEBI:18280
2-oxo-2-phenylacetic acid
OC(=O)C(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60°C to 69°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCO2H
Quantity 5 g
Beilstein 10,654
Solubility Information Solubility in water: .. Other solubilities: 1g/10mL methanol
Formula Weight 150.13
Percent Purity 97%
Physical Form Adhering Crystals, Crystalline Powder or Chunks
Chemical Name or Material Benzoylformic acid
Show More Show Less
compliance-icons
Product Identifier
  • Benzoylformic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'