missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Benzoylecgonine Monoclonal Antibody (3P1A6)

Product Code. 12857962
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12857962 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12857962 Supplier Invitrogen™ Supplier No. MIB0201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MIB0201 targets Benzoylecgonine in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIB0201 immunogen is benzoylecgonine-KLH. MIB0201 was formerly sold as a Seradyn product.

Benzoylecgonine is used as the main pharmaceutical ingredient in the prescription drug Esterom, a topical solution used for the relief of muscle pain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Benzoylecgonine
Applications ELISA, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 3P1A6
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias C16H19NO4
Host Species Mouse
Immunogen Benzoylecgonine-KLH.
Purification Method Protein A
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.