missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoylacetonitrile, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Benzoylacetonitrile |
| CAS | 614-16-4 |
| Molecular Formula | C9 H7 N O |
| Quantity | 10 g |
| Synonym | Benzenepropanenitrile, β-oxo-, Acetonitrile, benzoyl- (6CI,7CI,8CI), β-Oxobenzenepropanenitrile, 2-Cyanoacetophenone, 3-Oxo-3-phenylpropanenitrile, 3-Oxo-3-phenylpropionitrile, 3-Phenyl-3-ketopropionitrile, 3-Phenyl-3-oxopropanenitrile, Benzoylacetonitrile, Benzoylcyanomethane, Cyanomethyl phenyl ketone, NSC 4713, Phenacyl cyanide, α-Cyanoacetophenone, β-Oxobenzenepropionitrile, ω-Cyanoacetophenone |
| InChI Formula | InChI=1S/C9H7NO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5H,6H2 |
| SMILES | O=C(CC#N)c1ccccc1 |
| IUPAC Name | 3-oxo-3-phenylpropanenitrile |
| Molecular Weight (g/mol) | 145.16 |
| Formula Weight | 145.0528 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction