missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetonitrile, 98+%

Product Code. p-64215 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10599462

Brand: Thermo Scientific Acros 402000050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-16-4 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-16-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001942
InChI Key ZJRCIQAMTAINCB-UHFFFAOYSA-N
Synonym benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile
PubChem CID 64799
ChEBI CHEBI:51855
IUPAC Name 3-oxo-3-phenylpropanenitrile
SMILES O=C(CC#N)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 78.0°C to 82.0°C
Boiling Point 160.0°C (10.0 mmHg)
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5COCH2CN
Quantity 5 g
Beilstein 10, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 145.16
Percent Purity 98+%
Chemical Name or Material Benzoylacetonitrile, ≥98%
Show More Show Less
compliance-icons
Product Identifier
  • Benzoylacetonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.