missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetonitrile, 98+%

Product Code. 11491798
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491798 100 g 100g
11471798 5 g 5g
11481798 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491798 Supplier Thermo Scientific Alfa Aesar Supplier No. A14473.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 614-16-4 | C9H7NO | 145.16 g/mol

Benzoylacetonitrile is used to produce 2-benzoyl-3-furan-2-yl-acrylonitrile. It is also used in the synthesis of substituted naphtho[1,8-bc]pyrans and as building block in the preparation of 4H-pyrans, 2-pyridones, furans and carbocyclics. It is an active methylene compound, useful in heterocyclic synthesis, e.g. polyfunctional pyridines and pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoylacetonitrile is used to produce 2-benzoyl-3-furan-2-yl-acrylonitrile. It is also used in the synthesis of substituted naphtho[1,8-bc]pyrans and as building block in the preparation of 4H-pyrans, 2-pyridones, furans and carbocyclics. It is an active methylene compound, useful in heterocyclic synthesis, e.g. polyfunctional pyridines and pyrimidines.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-16-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001942
InChI Key ZJRCIQAMTAINCB-UHFFFAOYSA-N
Synonym benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile
PubChem CID 64799
ChEBI CHEBI:51855
IUPAC Name 3-oxo-3-phenylpropanenitrile
SMILES O=C(CC#N)C1=CC=CC=C1

Specifications

Melting Point 79°C to 83°C
Boiling Point 159°C to 160°C (10 mmHg)
Quantity 100 g
Beilstein 386745
Solubility Information Slightly soluble in water.
Formula Weight 145.16
Percent Purity ≥98%
Chemical Name or Material Benzoylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.