missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetone, 98+%

Product Code. 11411908
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411908 50 g 50g
11421908 250 g 250g
11401908 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411908 Supplier Thermo Scientific Alfa Aesar Supplier No. A14537.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-91-4 | C10H10O2 | 162.188 g/mol

Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoylacetone has been used as a 1,3-dicarbonyl compound model for studying keto-enol equilibria in aqueous acid and micellar solutions. Additionally, it has been studied using gas-phase electron diffraction and quantum chemistry. It is used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1

Specifications

Melting Point 55°C to 60°C
Density 1.09
Boiling Point 260°C to 262°C
Quantity 50 g
Beilstein 742413
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity ≥98%
Chemical Name or Material Benzoylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.