missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isothiocyanate, 98%

Product Code. 11317076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317076 5 g 5g
11327076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317076 Supplier Thermo Scientific Alfa Aesar Supplier No. L06964.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 532-55-8 | C8H5NOS | 163.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-55-8
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD00004815
InChI Key CPEKAXYCDKETEN-UHFFFAOYSA-N
Synonym benzoylthiocarbimide, benzoylisothiocyanate, benzoylisothiocyante, isothiocyanic acid benzoyl ester, unii-u8j7q32b86, n-benzoyl isothiocyanate, benzoic acid, anhydride with hncs, benzoic acid, anhydride with isothiocyanic acid, oxophenylmethanisothiocyanate, 1-benzoyl acetone
PubChem CID 68284
IUPAC Name benzoyl isothiocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=S

Specifications

Density 1.214
Boiling Point 128°C to 131°C (15 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6375
Quantity 5 g
UN Number UN2922
Beilstein 606716
Sensitivity Moisture sensitive
Merck Index 14,1114
Formula Weight 163.2
Percent Purity 98%
Chemical Name or Material Benzoyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzoyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzoyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.