missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl fluoride, 97%

Product Code. 11387786
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387786 25 g 25g
11377786 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387786 Supplier Thermo Scientific Alfa Aesar Supplier No. L11464.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-32-3 | C7H5FO | 124.11 g/mol

Benzoyl fluoride is used as a mild fluorinating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl fluoride is used as a mild fluorinating agent.

Solubility
Hydrolyses in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point ∼-28°C
Density 1.14
Boiling Point 159°C to 161°C
Flash Point 48°C (118°F)
Odor Pungent
Refractive Index 1.496
Quantity 25 g
UN Number UN2920
Beilstein 1904977
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 124.12
Percent Purity 98%
Chemical Name or Material Benzoyl fluoride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.