missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl fluoride, 97%

Product Code. 10612952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10612952 5 g 5g
15412152 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10612952 Supplier Thermo Scientific Acros Supplier No. 402040050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 455-32-3 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point -28.0°C
Density 1.1500g/mL
Boiling Point 159.0°C to 161.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COF
Refractive Index 1.496
Quantity 5 g
Beilstein 09,181
Specific Gravity 1.15
Solubility Information Solubility in water: hydrolyses.
Formula Weight 124.11
Percent Purity 97%
Chemical Name or Material Benzoyl fluoride
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Benzoyl fluoride
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P240-Ground/bond container and receiving equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P390-Absorb spillage to prevent material damage.
  • P402-Store in a dry place.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.