missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Cyanide, TRC

Product Code. 30332726
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30332726 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332726 Supplier Toronto Research Chemicals Supplier No. TRCB13037510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoyl Cyanide
CAS 613-90-1
Molecular Formula C8H5NO
Quantity 10 g
InChI Formula InChI=1S/C8H5NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5H
SMILES O=C(C#N)c1ccccc1
IUPAC Name benzoyl cyanide
Molecular Weight (g/mol) 131.13
Formula Weight 131.0371
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.