missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Chloride, TRC

Product Code. 30012244
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012244 500 mL 500mL
30007107 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012244 Supplier Toronto Research Chemicals Supplier No. TRCB209550500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7 H5 Cl O
Molecular Weight (g/mol) 140.57
Synonym Benzoyl Chloride, 7-Cyanoheptyl benzoate, B 0105, Benzaldehyde, α-chloro-, Benzenecarbonyl chloride, Benzoic acid chloride, Phenylcarbonyl chloride, Phenylcarboxyl chloride
IUPAC Name benzoyl chloride
SMILES ClC(=O)c1ccccc1

Specifications

InChI Formula InChI=1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H
Formula Weight 140.0029
Quantity 500 mL
Chemical Name or Material Benzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.