missing translation for 'onlineSavingsMsg'
Learn More

Benzoxazolemethanesulfonamide-N-(6-methyl-hexanoate), TRC

Product Code. 30336086
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336086 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336086 Supplier Toronto Research Chemicals Supplier No. TRCB2077305MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzoxazolemethanesulfonamide-N-(6-methyl-hexanoate)
CAS 1076198-89-4
Molecular Formula C15 H20 N2 O5 S
Quantity 5 mg
Synonym Hexanoic acid, 6-[[(2-benzoxazolylmethyl)sulfonyl]amino]-, methyl ester, Benzoxazolemethanesulfonamide-N-(6-methyl-hexanoate)
InChI Formula InChI=1S/C15H20N2O5S/c1-21-15(18)9-3-2-6-10-16-23(19,20)11-14-17-12-7-4-5-8-13(12)22-14/h4-5,7-8,16H,2-3,6,9-11H2,1H3
SMILES COC(=O)CCCCCNS(=O)(=O)Cc1oc2ccccc2n1
IUPAC Name methyl 6-(1,3-benzoxazol-2-ylmethylsulfonylamino)hexanoate
Molecular Weight (g/mol) 340.39
Formula Weight 340.1093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.