missing translation for 'onlineSavingsMsg'
Learn More

Benzovindiflupyr, TRC

Product Code. 30332059
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332059 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332059 Supplier Toronto Research Chemicals Supplier No. TRCB2069655MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzovindiflupyr
CAS 1072957-71-1
Purity Grade Notes HPLC
Molecular Formula C18 H15 Cl2 F2 N3 O
Quantity 5 mg
Synonym N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide, Benzovindiflupyr, N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazol-4-carboxamide, Solatenol
InChI Formula InChI=1S/C18H15Cl2F2N3O/c1-25-7-11(15(24-25)17(21)22)18(26)23-12-4-2-3-8-9-5-6-10(13(8)12)14(9)16(19)20/h2-4,7,9-10,17H,5-6H2,1H3,(H,23,26)
SMILES Cn1cc(C(=O)Nc2cccc3C4CCC(C4=C(Cl)Cl)c23)c(n1)C(F)F
IUPAC Name N-[11-(dichloromethylene)-3-tricyclo[6.2.1.0^{2,7}]undeca-2,4,6-trienyl]-3-(difluoromethyl)-1-methyl-pyrazole-4-carboxamide
Molecular Weight (g/mol) 398.234
Formula Weight 397.056
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.