missing translation for 'onlineSavingsMsg'
Learn More

Benzovindiflupyr, TRC

Product Code. 18043630
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043630 2.5 mg 2.5mg
18042030 10 mg 10mg
18042830 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18043630 Supplier Toronto Research Chemicals Supplier No. TRCB2069652.5MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072957-71-1
Molecular Formula C18 H15 Cl2 F2 N3 O
Molecular Weight (g/mol) 398.234
Synonym N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazole-4-carboxamide, Benzovindiflupyr, N-[9-(Dichloromethylene)-1,2,3,4-tetrahydro-1,4-methanonaphthalen-5-yl]-3-(difluoromethyl)-1-methyl-1H-pyrazol-4-carboxamide, Solatenol
IUPAC Name N-[11-(dichloromethylene)-3-tricyclo[6.2.1.0^{2,7}]undeca-2,4,6-trienyl]-3-(difluoromethyl)-1-methyl-pyrazole-4-carboxamide
SMILES Cn1cc(C(=O)Nc2cccc3C4CCC(C4=C(Cl)Cl)c23)c(n1)C(F)F

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C18H15Cl2F2N3O/c1-25-7-11(15(24-25)17(21)22)18(26)23-12-4-2-3-8-9-5-6-10(13(8)12)14(9)16(19)20/h2-4,7,9-10,17H,5-6H2,1H3,(H,23,26)
Formula Weight 397.056
Percent Purity >95%
Chemical Name or Material Benzovindiflupyr
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.