missing translation for 'onlineSavingsMsg'
Learn More

Benzotropine Mesylate, TRC

Product Code. 30136365
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136365 50 mg 50mg
30135022 100 mg 100mg
30040360 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136365 Supplier Toronto Research Chemicals Supplier No. TRCB20757550MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-17-2
Molecular Formula C21 H25 N O . C H4 O3 S
Molecular Weight (g/mol) 403.54
Synonym 1αH,5αH-Tropane, 3α-(diphenylmethoxy)-, methanesulfonate, 8-Azabicyclo[3.2.1]octane, 3-(diphenylmethoxy)-8-methyl-, (3-endo)-, methanesulfonate, 8-Azabicyclo[3.2.1]octane, 3-(diphenylmethoxy)-8-methyl-, endo-, methanesulfonate, 3-Diphenylmethoxytropane mesylate, 3-Diphenylmethoxytropane methanesulfonate, Benzatropine methanesulfonate, Benzotropine mesylate, Benzotropine methanesulfonate, Benztropine mesylate, Benztropine methanesulfonate, Cobrentin methanesulfonate, Cogentin, Cogentin methanesulfonate, Cogentinol, MK 02, NSC 169913, Tropine benzohydryl ether methanesulfonate
IUPAC Name (1R,5S)-3-benzhydryloxy-8-methyl-8-azabicyclo[3.2.1]octane;methanesulfonic acid
SMILES CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(c3ccccc3)c4ccccc4.CS(=O)(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C21H25NO.CH4O3S/c1-22-18-12-13-19(22)15-20(14-18)23-21(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-5(2,3)4/h2-11,18-21H,12-15H2,1H3;1H3,(H,2,3,4)/t18-,19+,20+;
Formula Weight 403.1817
Chemical Name or Material Benztropine Mesilate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.