missing translation for 'onlineSavingsMsg'
Learn More

Benzotriazole BT, TRC

Product Code. 30138566
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138566 100 g 100g
17979374 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138566 Supplier Toronto Research Chemicals Supplier No. TRCB206990100G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70321-86-7
Molecular Formula C30 H29 N3 O
Molecular Weight (g/mol) 447.57
Synonym 2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol, 2-(2-Hydroxy-3,5-di-α-cumylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-dicumyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-l-phenylethyl)phenol, 2-(2H-Benzotriazol-2-yl)-4,6-bis(α,α-dimethylbenzyl)phenol, 2-(3',5'-Bis(α,α-dimethylbenzyl)-2'-hydroxyphenyl)benzotriazole, 2-(Benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol, 2-[2-Hydroxy-3,5-bis(α,α-dimethylbenzyl)phenyl]-2H-benzotriazole, 2-[2-Hydroxy-3,5-bis(α,α-dimethylbenzyl)phenyl]benzotriazole, 2-[2-Hydroxy-3,5-di(1,1-dimethylbenzyl)phenyl]-2H-benzotriazole, 2-[2-Hydroxy-3,5-di(α,α-dimethylbenzyl)phenyl]-2H-benzotriazole, 2-[2-Hydroxy-3,5-di-( 1,1-dimethylbenzyl)]-2H-benzotriazole, 2-[2'-Hydroxy-3',5'-bis(α,α-dimethylbenzyl)phenyl]benzotriazole, 2-[3',5'-Bis(1-methyl-1-phenylethyl)-2'-hydroxyphenyl]benzotriazole, BLS 234, Benzotriazole BT, Chiguard 234, Chisorb 234, Eversorb 234, Eversorb 76, Jinwei 234, Light Stabilizer 234, Lowilite 234, Milestab 234PD, Songsorb 2340, T 234, Tinuvin 234, Tinuvin 234D, Tinuvin 900, UV 234, UV 234 (antioxidant), UVA 234, Uvinul 3034
IUPAC Name 2-(benzotriazol-2-yl)-4,6-bis(2-phenylpropan-2-yl)phenol
SMILES CC(C)(c1ccccc1)c2cc(c(O)c(c2)C(C)(C)c3ccccc3)n4nc5ccccc5n4

Specifications

Quantity 100 g
InChI Formula InChI=1S/C30H29N3O/c1-29(2,21-13-7-5-8-14-21)23-19-24(30(3,4)22-15-9-6-10-16-22)28(34)27(20-23)33-31-25-17-11-12-18-26(25)32-33/h5-20,34H,1-4H3
Formula Weight 447.2311
Chemical Name or Material 2-(2H-Benzotriazol-2-yl)-4,6-bis(1-methyl-1-phenylethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.