missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, TRC

Product Code. 18050430
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050430 1 g 1g
18062220 5 g 5g
18080830 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18050430 Supplier Toronto Research Chemicals Supplier No. TRCB2066401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.19
Synonym Benzothiazole, 1,3-Benzothiazole, 1-Thia-3-azaindene, Benzo[d]thiazole, Benzosulfonazole, NSC 8040, Vangard BT
IUPAC Name 1,3-benzothiazole
SMILES c1ccc2scnc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H5NS/c1-2-4-7-6(3-1)8-5-9-7/h1-5H
Formula Weight 135.0143
Percent Purity >95%
Chemical Name or Material Benzothiazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.