missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, 97%

Product Code. 10570984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10570984 2.5 L Glass bottle 2.5 L
10436970 5 mL Glass bottle 5mL
10446970 250 mL Glass bottle 250mL
10133000 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10570984 Supplier Thermo Scientific Acros Supplier No. 146390025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-16-9 | C7H5NS | 135.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00005775
InChI Key IOJUPLGTWVMSFF-UHFFFAOYSA-N
Synonym benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
PubChem CID 7222
ChEBI CHEBI:45993
IUPAC Name 1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=CS2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 2°C
Color Brown to Brown-Yellow
Density 1.2300g/mL
Boiling Point 231°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6410 to 1.644
Quantity 2.5 L
Fieser 15,20
Merck Index 15, 1110
Specific Gravity 1.23
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and carbon disulfide
Formula Weight 135.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzothiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.