missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, 97%

Product Code. 11454270
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11454270

Brand: Thermo Scientific Alfa Aesar A10380.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-16-9 | C7H5NS | 135.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is an interesting carbonyl equivalent and reacts with aldehydes or ketones to generate α-hydroxy carbonyl compounds. Benzothiazole is used as a chemical intermediate in organic synthesis. It is a precursor of rubber accelerators and a component of cyanine dyes. It is also used as a flavoring substance. It has been used as an antimicrobial agent in athletic shoe insoles. It is also used as a flavor, antimicrobial agent, and component of cyanine dyes. Its derivatives are used as rubber accelerators.

Solubility
Slightly soluble in water. Soluble in alcohol, carbon disulfide

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.184
MDL Number MFCD00005775
InChI Key IOJUPLGTWVMSFF-UHFFFAOYSA-N
Synonym benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
PubChem CID 7222
ChEBI CHEBI:45993
IUPAC Name 1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=CS2

Specifications

Melting Point 2°C
Density 1.241
Boiling Point 230°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.642
Quantity 250 g
UN Number UN2810
Beilstein 109468
Merck Index 14,1107
Solubility Information Slightly soluble in water; Soluble in alcohol, carbon disulfide
Formula Weight 135.19
Percent Purity 97%
Chemical Name or Material Benzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • Benzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Benzothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.