missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, 97%

Product Code. 11444270
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444270 50 g 50g
11454270 250 g 250g
11434270 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444270 Supplier Thermo Scientific Alfa Aesar Supplier No. A10380.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-16-9 | C7H5NS | 135.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190-254nm range, to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L)
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.184
MDL Number MFCD00005775
InChI Key IOJUPLGTWVMSFF-UHFFFAOYSA-N
Synonym benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
PubChem CID 7222
ChEBI CHEBI:45993
IUPAC Name 1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=CS2

Specifications

Melting Point 2°C
Density 1.241
Boiling Point 230°C
Flash Point 107°C (224°F)
Odor Characteristic
Refractive Index 1.642
Quantity 50 g
UN Number UN2810
Beilstein 109468
Merck Index 14,1107
Solubility Information Slightly soluble in water; Soluble in alcohol, carbon disulfide
Formula Weight 135.19
Percent Purity 97%
Chemical Name or Material Benzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • Benzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Benzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.