missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole-6-carboxylic acid, 96%

Product Code. 15434527
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434527 1 g 1g
15444527 5 g 5g
15454527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15434527 Supplier Thermo Scientific Alfa Aesar Supplier No. H31868.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 3622-35-3 | C8H5NO2S | 179.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3622-35-3
Molecular Formula C8H5NO2S
Molecular Weight (g/mol) 179.19
MDL Number MFCD00111651
InChI Key DMPZJACLHDWUFS-UHFFFAOYSA-N
Synonym benzothiazole-6-carboxylic acid, 6-benzothiazolecarboxylic acid, benzo d thiazole-6-carboxylic acid, zlchem 73, pubchem9871, enamine_005417, benzothiazole-6-carboxylic, 6-benzothiazolecarboxylicacid, 6-benzothiazole carboxylic acid, cl-cob-iii-274-2
PubChem CID 601670
IUPAC Name 1,3-benzothiazole-6-carboxylic acid
SMILES OC(=O)C1=CC=C2N=CSC2=C1

Specifications

Melting Point 245°C to 251°C
Quantity 1 g
Formula Weight 179.2
Percent Purity 96%
Chemical Name or Material Benzothiazole-6-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.