missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoquinonium Chloride, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Benzoquinonium Chloride |
| CAS | 311-09-1 |
| Analyte or Component Names | Benzoquinonium Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34 H50 N4 O2 . 2 Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ammonium, [p-benzoquinon-2,5-ylenebis(iminotrimethylene)]bis[benzyldiethyl-, dichloride (8CI), Benzenemethanaminium, N,N'-[(3,6-dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethyl-, dichloride (9CI), [p-Benzoquinon-2,5-ylenebis(iminotrimethylene)]bis[benzyldiethylammonium chloride] (6CI,7CI), 2,5-Bis(3-diethylaminopropylamino)benzoquinone bis(benzyl chloride), Amilyt, Benzoquinonium chloride, Mytolon, Win 2747, Benzenemethanaminium, N,N'-[(3,6-dioxo-1,4-cyclohexadiene-1,4-diyl)bis(imino-3,1-propanediyl)]bis[N,N-diethyl-, chloride (1:2) |
| InChI Formula | InChI=1S/C34H48N4O2.2ClH/c1-5-37(6-2,27-29-17-11-9-12-18-29)23-15-21-35-31-25-34(40)32(26-33(31)39)36-22-16-24-38(7-3,8-4)28-30-19-13-10-14-20-30;;/h9-14,17-20,25-26H,5-8,15-16,21-24,27-28H2,1-4H3;2*1H |
| Solution Type | Neat |
| SMILES | [Cl-].[Cl-].CC[N+](CC)(CCCNC1=CC(=O)C(=CC1=O)NCCC[N+](CC)(CC)Cc2ccccc2)Cc3ccccc3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?