missing translation for 'onlineSavingsMsg'
Learn More

Benzopurpurin 4B

Product Code. 15486629
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15486629

Brand: Thermo Scientific Alfa Aesar J64901.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 992-59-6 | C34H26N6Na2O6S2 | 724.718 g/mol

Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzopurpurine 4B is a biological stain and pH indicator for pH 2.3-4.4.

Solubility
Soluble in water (20 mg/ml), acetone, ethanol (3 mg/ml), sodium hydroxide, and sulfuric acid.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 992-59-6
Molecular Formula C34H26N6Na2O6S2
Molecular Weight (g/mol) 724.718
MDL Number MFCD00012410
InChI Key SUXCALIDMIIJCK-UHFFFAOYSA-L
Synonym benzopurpurine 4b, benzopurpurin 4b, direct red 2, c.i. direct red 2, amanil purpurine, diphenyl red 4b, hispamin red 4b, direct red dcb, azocard red 4b, diazine red 4b
PubChem CID 13816
IUPAC Name disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]-3-methylphenyl]-2-methylphenyl]diazenyl]naphthalene-1-sulfonate
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=NC3=C(C4=CC=CC=C4C(=C3)S(=O)(=O)[O-])N)C)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N.[Na+].[Na+]

Specifications

Color Red
Quantity 100 g
Merck Index 14,1011
Solubility Information Soluble in water (20mg/ml),acetone,ethanol (3mg/ml),sodium hydroxide,and sulfuric acid.
Formula Weight 724.73
Physical Form Powder
Chemical Name or Material Benzopurpurin 4B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.