missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacole, 98%, Thermo Scientific Chemicals

Product Code. 10418630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418630 100 g Plastic bottle 100g
10559630 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10418630 Supplier Thermo Scientific Acros Supplier No. 105591000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 464-72-2
Molecular Formula C26H22O2
Molecular Weight (g/mol) 366.45
MDL Number MFCD00004448
InChI Key MFEWNFVBWPABCX-UHFFFAOYSA-N
Synonym benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
PubChem CID 94766
IUPAC Name 1,1,2,2-tetraphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 184°C to 186°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (C6H5)2C(OH)C(OH)(C6H5)2
Quantity 100 g
Beilstein 06, 1058
Merck Index 15, 1103
Solubility Information Solubility in water: insoluble
Formula Weight 366.45
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Benzopinacole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.