missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzopinacole, 98%, Thermo Scientific Chemicals
£17.40 - £86.00
Chemical Identifiers
CAS | 464-72-2 |
---|---|
Molecular Formula | C26H22O2 |
Molecular Weight (g/mol) | 366.45 |
MDL Number | MFCD00004448 |
InChI Key | MFEWNFVBWPABCX-UHFFFAOYSA-N |
Synonym | benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl |
PubChem CID | 94766 |
IUPAC Name | 1,1,2,2-tetraphenylethane-1,2-diol |
SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10559630
|
Thermo Scientific Acros
105590100 |
10 g | Glass bottle |
£17.40
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10418630
|
Thermo Scientific Acros
105591000 |
100 g | Plastic bottle |
£86.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
464-72-2 | |
366.45 | |
MFEWNFVBWPABCX-UHFFFAOYSA-N | |
94766 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O |
C26H22O2 | |
MFCD00004448 | |
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl | |
1,1,2,2-tetraphenylethane-1,2-diol |
Specifications
464-72-2 | |
100.0 | |
White to Yellow | |
97.5% min. (HPLC) | |
(C6H5)2C(OH)C(OH)(C6H5)2 | |
06, 1058 | |
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl | |
MFEWNFVBWPABCX-UHFFFAOYSA-N | |
1,1,2,2-tetraphenylethane-1,2-diol | |
94766 | |
98% | |
Benzopinacole, 98% |
97.5 | |
184°C to 186°C | |
Authentic | |
C26H22O2 | |
MFCD00004448 | |
15, 1103 | |
Solubility in water: insoluble | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O | |
366.45 | |
366.45 | |
Crystals or Powder |
Safety and Handling
missing translation for 'einecsNumber' : 207-356-8
missing translation for 'rtecsNumber' : KI2750000
missing translation for 'tsca' : TSCA
RUO – Research Use Only