missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzopinacol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications: Benzopinacol is a catalyst of the formation of unsaturated polyesters. It is also used as an initiator of polymerisation by free radicals. It is also used as an organic synthesis intermediate.
Specifications
Melting Point | 174°C to 180°C |
Quantity | 25 g |
Beilstein | 2061947 |
Merck Index | 14,1100 |
Solubility Information | It is soluble in ethanol,ether,acetone insoluble in carbon disulfide and water. |
Formula Weight | 366.46 |
Percent Purity | 98% |
Chemical Name or Material | Benzopinacol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'