missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone hydrazone, 98+%

Product Code. 11494973
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494973 50 g 50g
11404983 250 g 250g
11484973 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494973 Supplier Thermo Scientific Alfa Aesar Supplier No. A12266.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5350-57-2 | C13H12N2 | 196.253 g/mol

An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An important compound in organic photochemistry and perfumery as well as in organic synthesis. Used in the antibiotics synthesis of 6-APA and 7-ACA of the carboxyl protecting groups and other organic compounds. Used as organic pigment and medical intermediate. It is used as a photoinitiator of UV-curing applications in inks, adhesive, coatings and optical fiber.

Solubility
Soluble in ether, benzene, chloroform and other organic solvents.

Notes
Store in a tightly closed container. Store in a cool, dry, well-ventilated area away from incompatible substances. Keep refrigerated. (Store below 4 C/39 F.)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5350-57-2
Molecular Formula C13H12N2
Molecular Weight (g/mol) 196.253
MDL Number MFCD00007624
InChI Key QYCSNMDOZNUZIT-UHFFFAOYSA-N
Synonym benzophenone hydrazone, diphenylmethylene hydrazine, benzophenonehydrazone, methanone, diphenyl-, hydrazone, diphenylmethanone hydrazone, benzophenone, hydrazone, diphenylmethylidene hydrazine, diphenyl ketone hydrazone, benzophenone hydrozone, nsc 43
PubChem CID 79304
IUPAC Name benzhydrylidenehydrazine
SMILES C1=CC=C(C=C1)C(=NN)C2=CC=CC=C2

Specifications

Melting Point 96°C to 100°C
Density 1.1
Boiling Point 225°C to 230°C (55 mmHg)
Flash Point 146°C (294°F)
Odor Characteristic
Quantity 50 g
UN Number UN2811
Beilstein 1910177
Solubility Information Soluble in ether,benzene,chloroform and other organic solvents.
Formula Weight 196.25
Percent Purity ≥98%
Chemical Name or Material Benzophenone hydrazone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.