missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone, 99%, pure

Product Code. 10627632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10627632 1 kg Plastic bottle 1kg
10529980 500 g Plastic bottle 500g
11974841 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10627632 Supplier Thermo Scientific Acros Supplier No. 105560010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-61-9 | C13H10O | 182.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-61-9
Molecular Formula C13H10O
Molecular Weight (g/mol) 182.22
InChI Key RWCCWEUUXYIKHB-UHFFFAOYSA-N
Synonym benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
PubChem CID 3102
ChEBI CHEBI:41308
IUPAC Name diphenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 47.0°C to 49.0°C
Color White
Boiling Point 305.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)2CO
Quantity 1 kg
Beilstein 07,411
Fieser 13,108
Merck Index 15,11
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene
Formula Weight 182.22
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Benzophenone
Show More Show Less
compliance-icons
Produktbeteckning
  • Benzophenone
Signalord
  • Danger
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Faroangivelser
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Skyddsangivelse
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.