missing translation for 'onlineSavingsMsg'
Learn More

Benzonitrile, TRC

Product Code. 30337128
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30337128 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30337128

Brand: Toronto Research Chemicals TRCB2046005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzonitrile
CAS 100-47-0
Molecular Formula C7 H5 N
Quantity 5 g
Synonym Benzene, cyano-, Benzenecarbonitrile, Benzenenitrile, Benzoic acid nitrile, Benzonitril, Cyanobenzene, NSC 8039, Phenyl cyanide, Benzonitrile
InChI Formula InChI=1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H
SMILES N#Cc1ccccc1
IUPAC Name benzonitrile
Molecular Weight (g/mol) 103.12
Formula Weight 103.0422
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.